Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015164.2 | NP_055979.2 | 2 | 303 | ||
XM_005245790.3 | XP_005245847.1 | 2 | 498 | |||
XM_017000757.1 | XP_016856246.1 | 2 | 210 | |||
XM_017000758.1 | XP_016856247.1 | 2 | 210 | |||
GenBank mRNA | AB020649.1 | - | 2 | 79 | ||
BC040441.1 | - | 2 | 306 |