Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: U2SURP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080415.1 | NP_001073884.1 | 3 | 295 | ||
NM_001320219.1 | NP_001307148.1 | 3 | 295 | |||
NM_001320220.1 | NP_001307149.1 | 3 | 295 | |||
NM_001320222.1 | NP_001307151.1 | 3 | 295 | |||
XM_005247248.4 | XP_005247305.1 | 3 | 301 | |||
XM_017006038.1 | XP_016861527.1 | 3 | 234 | |||
GenBank mRNA | AB002330.1 | - | 3 | 194 | ||
AK296440.1 | - | 3 | 215 | |||
BC006474.1 | - | 3 | 194 | |||
BC016323.1 | - | 3 | 212 | |||
BC062727.1 | - | 4 | 312 | |||
BC105604.1 | - | 3 | 227 | |||
BC111692.1 | - | 3 | 666 | 40028600 |