Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDCA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005273733.3 | XP_005273790.1 | 8 | 1068 | ||
XM_005273734.3 | XP_005273791.1 | 8 | 1068 | |||
XM_011544743.2 | XP_011543045.1 | 8 | 1068 | |||
XM_011544747.2 | XP_011543049.1 | 7 | 624 | |||
XM_011544748.2 | XP_011543050.1 | 8 | 1093 | |||
XM_017017158.1 | XP_016872647.1 | 8 | 1068 |