Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012166.2 | NP_036298.2 | 2 | 563 | ||
XM_005251439.4 | XP_005251496.1 | 2 | 652 | |||
XM_017014618.1 | XP_016870107.1 | 2 | 769 | |||
XM_017014619.1 | XP_016870108.1 | 2 | 880 | |||
GenBank mRNA | AF176705.1 | - | 2 | 562 | ||
BC125125.1 | - | 2 | 642 | 40119019 | ||
BC140785.1 | - | 1 | 534 | 9021653 | ||
BC171785.1 | - | 1 | 534 | 9054439 | ||
CN411883.1 | - | 2 | 563 |