Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C17orf89 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001086521.1 | NP_001079990.1 | 3 | 383 | ||
GenBank mRNA | AA534550.1 | - | 2 | 202 | ||
BC079831.1 | - | 3 | 374 |