Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMSD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001123366.1 | NP_001116838.1 | 4 | 581 | ||
GenBank mRNA | BM696260.1 | - | 4 | 581 |