Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC22A10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039752.3 | NP_001034841.3 | 6 | 1207 | ||
NR_134874.1 | 7 | 962 | ||||
XM_011545016.2 | XP_011543318.1 | 8 | 2212 | |||
XM_017017699.1 | XP_016873188.1 | 7 | 1009 | |||
XM_017017700.1 | XP_016873189.1 | 6 | 1445 | |||
XM_017017701.1 | XP_016873190.1 | 8 | 2212 | |||
XM_017017702.1 | XP_016873191.1 | 8 | 2212 | |||
XM_017017703.1 | XP_016873192.1 | 5 | 553 | |||
XM_017017704.1 | XP_016873193.1 | 6 | 1444 | |||
GenBank mRNA | AB075876.1 | - | 7 | 962 | ||
AK296299.1 | - | 8 | 914 | |||
AK298430.1 | - | 7 | 1009 | |||
BC171744.1 | - | 4 | 542 |