Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161330.1 | NP_001154802.1 | 3, 4 | 309 | ||
NM_001161331.1 | NP_001154803.1 | 2, 3 | 401 | |||
NM_018984.3 | NP_061857.3 | 3, 4 | 309 | |||
XM_005268984.4 | XP_005269041.1 | 3, 4 | 1020 | |||
XM_005268985.2 | XP_005269042.1 | 2, 3 | 135 | |||
XM_011538497.1 | XP_011536799.1 | 2, 3 | 401 | |||
XM_011538499.1 | XP_011536801.1 | 2, 3 | 401 | |||
XM_017019491.1 | XP_016874980.1 | 2, 3 | 1247 | |||
XM_017019492.1 | XP_016874981.1 | 2, 3 | 291 | |||
XM_017019493.1 | XP_016874982.1 | 2, 3 | 153 | |||
XM_017019494.1 | XP_016874983.1 | 2, 3 | 248 | |||
GenBank mRNA | AB072355.1 | - | 3, 4 | 222 | ||
AB072356.1 | - | 3, 4 | 222 | |||
AB072357.1 | - | 1 | 96 | |||
AK095421.1 | - | 2, 3 | 402 | |||
BC062341.1 | - | 3, 4 | 301 | 6376684 | ||
BP363810.1 | - | 3, 4 | 309 |