Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257158.1 | NP_001244087.1 | 5, 6 | 521 | ||
NM_001257159.1 | NP_001244088.1 | 4, 5 | 473 | |||
NM_018718.2 | NP_061188.1 | 5, 6 | 521 | |||
NR_046443.1 | 5, 6 | 521 | ||||
XM_011516709.2 | XP_011515011.1 | 6, 7 | 482 | |||
XM_011516710.2 | XP_011515012.1 | 4, 5 | 297 | |||
GenBank mRNA | AF429308.1 | - | 5, 6 | 367 | ||
AJ278890.1 | - | 5, 6 | 324 | |||
AK026098.1 | - | 5, 6 | 321 | |||
AK298618.1 | - | 4, 5 | 289 | |||
AK311382.1 | - | 5, 6 | 338 | |||
AK314676.1 | - | 5, 6 | 300 | |||
BC018789.1 | - | 5, 6 | 303 | |||
BC056162.1 | - | 5, 6 | 294 | 6142659 | ||
DB069500.1 | - | 523 |