Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRANK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014831.2 | NP_055646.2 | 6, 7 | 896 | ||
XM_006713432.3 | XP_006713495.1 | 7, 8 | 1064 | |||
XM_011534289.2 | XP_011532591.1 | 7, 8 | 1825 | |||
XM_017007570.1 | XP_016863059.1 | 7, 8 | 867 | |||
XM_017007571.1 | XP_016863060.1 | 7, 8 | 957 | |||
XM_017007572.1 | XP_016863061.1 | 7, 8 | 859 | |||
XM_017007573.1 | XP_016863062.1 | 7, 8 | 867 | |||
GenBank mRNA | EG328357.1 | - | 4, 5 | 397 |