Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKUB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144960.2 | NP_001138432.1 | 4 | 1000 | ||
NM_001315505.1 | NP_001302434.1 | 4 | 890 | |||
NM_001315506.1 | NP_001302435.1 | 4 | 1000 | |||
XM_011512798.1 | XP_011511100.1 | 4 | 497 | |||
XM_011512799.1 | XP_011511101.1 | 4 | 731 | |||
XM_011512801.1 | XP_011511103.1 | 2 | 168 | |||
XM_011512802.1 | XP_011511104.1 | 4 | 731 | |||
GenBank mRNA | AK304359.1 | - | 4 | 731 | ||
BC171805.1 | - | 4 | 890 |