Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161330.1 | NP_001154802.1 | 4 | 332 | ||
NM_001161331.1 | NP_001154803.1 | 3 | 424 | |||
NM_018984.3 | NP_061857.3 | 4 | 332 | |||
XM_005268984.4 | XP_005269041.1 | 4 | 1043 | |||
XM_005268985.2 | XP_005269042.1 | 3 | 158 | |||
XM_011538497.1 | XP_011536799.1 | 3 | 424 | |||
XM_011538499.1 | XP_011536801.1 | 3 | 424 | |||
XM_011538500.2 | XP_011536802.1 | 3 | 248 | |||
XM_011538501.2 | XP_011536803.1 | 3 | 244 | |||
XM_017019491.1 | XP_016874980.1 | 3 | 1270 | |||
XM_017019492.1 | XP_016874981.1 | 3 | 314 | |||
XM_017019493.1 | XP_016874982.1 | 3 | 176 | |||
XM_017019494.1 | XP_016874983.1 | 3 | 271 | |||
GenBank mRNA | AB072355.1 | - | 4 | 245 | ||
AB072356.1 | - | 4 | 245 | |||
AB072357.1 | - | 1 | 119 | |||
AK095421.1 | - | 3 | 425 | |||
BC062341.1 | - | 4 | 324 | 6376684 | ||
BP363810.1 | - | 4 | 332 |