Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257158.1 | NP_001244087.1 | 10 | 2395 | ||
NM_001257159.1 | NP_001244088.1 | 9 | 2347 | |||
NM_018718.2 | NP_061188.1 | 11 | 2611 | |||
NR_046443.1 | 11 | 2535 | ||||
XM_011516709.2 | XP_011515011.1 | 12 | 2572 | |||
XM_011516710.2 | XP_011515012.1 | 10 | 2387 | |||
GenBank mRNA | AF429308.1 | - | 11 | 2457 | ||
AJ278890.1 | - | 11 | 2414 | |||
AK026098.1 | - | 11 | 2411 | |||
AL359617.1 | - | 5 | 1912 | |||
AY186739.1 | - | 3 | 2053 | |||
BC018789.1 | - | 10 | 2177 | |||
BC047500.1 | - | 1 | 109 | |||
BC056162.1 | - | 10 | 2168 | 6142659 |