Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DTHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136536.4 | NP_001130008.2 | 2 | 647 | ||
NM_001170700.2 | NP_001164171.1 | 2 | 802 | |||
XM_006714014.3 | XP_006714077.1 | 3 | 1281 | |||
XM_011513693.2 | XP_011511995.1 | 3 | 1282 | |||
XM_011513694.2 | XP_011511996.1 | 3 | 1282 | |||
XM_011513695.2 | XP_011511997.1 | 3 | 1281 | |||
XM_011513696.2 | XP_011511998.1 | 2 | 665 | |||
XM_017008191.1 | XP_016863680.1 | 3 | 1282 | |||
XM_017008192.1 | XP_016863681.1 | 3 | 1284 | |||
XM_017008193.1 | XP_016863682.1 | 3 | 1283 | |||
GenBank mRNA | AK131492.1 | - | 2 | 640 | ||
AK304357.1 | - | 2 | 633 | |||
AK316523.1 | - | 2 | 633 | |||
BC157885.1 | - | 2 | 802 | 9057096 |