Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRRT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114726.2 | NP_001108198.2 | 3 | 933 | ||
NM_001174164.1 | NP_001167635.1 | 4 | 1050 | |||
XM_005250342.3 | XP_005250399.1 | 3 | 1050 | |||
XM_005250343.3 | XP_005250400.1 | 3 | 959 | |||
XM_006715985.3 | XP_006716048.1 | 3 | 1063 | |||
XM_017012205.1 | XP_016867694.1 | 4 | 1869 | |||
XM_017012206.1 | XP_016867695.1 | 3 | 1842 | |||
GenBank mRNA | BX327107.2 | - | 2 | 215 |