Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM64A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019013.2 | NP_061886.2 | 5, 6 | 848 | ||
GenBank mRNA | AK001018.1 | - | 4, 5 | 788 | ||
AK225613.1 | - | 5, 6 | 810 | |||
AK225702.1 | - | 5, 6 | 808 | |||
BC013966.2 | - | 5, 6 | 794 | 2988318 | ||
KU178654.1 | - | 4, 5 | 739 |