Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHRS4L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC041992.1 | - | 1, 2 | 394 | ||
KC237374.1 | - | 1, 2 | 402 | |||
KU141377.1 | - | 1, 2 | 402 |