Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144952.1 | NP_001138424.1 | 12 | 1517 | ||
XM_011524914.2 | XP_011523216.1 | 12 | 1901 | |||
XM_011524915.2 | XP_011523217.1 | 12 | 1903 | |||
XM_011524916.2 | XP_011523218.1 | 12 | 1911 | |||
GenBank mRNA | AB040947.1 | - | 5 | 548 | ||
BC066363.1 | - | 4 | 417 |