Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ROBO4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301088.1 | NP_001288017.1 | 4 | 549 | ||
NM_019055.5 | NP_061928.4 | 4 | 755 | |||
XM_006718861.1 | XP_006718924.1 | 4 | 688 | |||
GenBank mRNA | AF361473.1 | - | 4 | 684 | ||
AK027852.1 | - | 4 | 654 | |||
AK074163.1 | - | 3 | 285 | |||
AK289769.1 | - | 4 | 654 | |||
AK302628.1 | - | 4 | 419 | |||
AY358083.1 | - | 4 | 606 | |||
BC014995.1 | - | 3 | 367 | |||
BC039602.1 | - | 4 | 643 | 5590503 | ||
BC111562.1 | - | 4 | 649 | 40068748 | ||
BC111748.1 | - | 4 | 649 | 40068752 |