Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXW5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018998.3 | NP_061871.1 | 9 | 1972 | ||
XM_005266089.2 | XP_005266146.2 | 9 | 2022 | |||
XM_011518789.1 | XP_011517091.1 | 9 | 1968 | |||
XM_017014812.1 | XP_016870301.1 | 9 | 1938 | |||
GenBank mRNA | AB209726.1 | - | 6 | 1384 | ||
AF217998.1 | - | 5 | 1131 | |||
AF258569.1 | - | 5 | 1131 | |||
AK026081.1 | - | 9 | 1836 | |||
AL137631.1 | - | 5 | 1526 | |||
BC000850.1 | - | 3 | 578 | |||
BC004541.2 | - | 6 | 1381 | |||
BC009429.1 | - | 4 | 1125 | |||
BC014130.1 | - | 9 | 1868 | 4620371 | ||
BC014297.2 | - | 9 | 1839 | 3958395 | ||
BC031100.1 | - | 8 | 1933 | |||
BC035355.1 | - | 8 | 1986 |