Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLXNA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020911.1 | NP_065962.1 | 16, 17 | 3386 | ||
XM_005250686.4 | XP_005250743.1 | 16, 17 | 3784 | |||
XM_006716171.3 | XP_006716234.1 | 17, 18 | 3877 | |||
XM_011516676.2 | XP_011514978.1 | 16, 17 | 3292 | |||
XM_017012779.1 | XP_016868268.1 | 15, 16 | 3091 | |||
GenBank mRNA | AB046770.3 | - | 14, 15 | 1863 |