Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MUC3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005960.1 | NP_005951.1 | 2 | 1190 | ||
XM_011516246.2 | XP_011514548.1 | 2 | 1549 | |||
XM_011516247.2 | XP_011514549.1 | 2 | 1550 | |||
XM_011516249.2 | XP_011514551.1 | 2 | 1554 | |||
XM_011516250.2 | XP_011514552.1 | 2 | 1549 | |||
XM_017012231.1 | XP_016867720.1 | 2 | 1556 |