Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021623.1 | NP_067636.1 | 9 | 964 | ||
XM_011544605.2 | XP_011542907.1 | 9 | 849 | |||
XM_011544606.2 | XP_011542908.1 | 9 | 900 | |||
XM_011544607.2 | XP_011542909.1 | 9 | 862 | |||
XM_017013718.1 | XP_016869207.1 | 8 | 715 | |||
XM_017013719.1 | XP_016869208.1 | 8 | 685 | |||
XM_017013720.1 | XP_016869209.1 | 8 | 800 | |||
GenBank mRNA | AK291842.1 | - | 9 | 908 | ||
AK296068.1 | - | 8 | 757 | |||
AK309020.1 | - | 2 | 81 | |||
DC332022.1 | - | 2 | 81 |