Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEAK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024776.3 | NP_079052.2 | 5 | 848 | ||
XM_005254670.2 | XP_005254727.1 | 8 | 1152 | |||
XM_005254671.2 | XP_005254728.1 | 7 | 1049 | |||
XM_005254673.2 | XP_005254730.1 | 5 | 783 | |||
XM_005254674.2 | XP_005254731.1 | 6 | 996 | |||
XM_005254675.4 | XP_005254732.1 | 2 | 502 | |||
XM_011522033.2 | XP_011520335.1 | 9 | 1208 | |||
XM_011522034.2 | XP_011520336.1 | 10 | 1316 | |||
XM_011522035.2 | XP_011520337.1 | 12 | 1467 | |||
XM_011522036.2 | XP_011520338.1 | 11 | 1371 | |||
XM_011522037.2 | XP_011520339.1 | 11 | 1412 | |||
XM_011522038.2 | XP_011520340.1 | 10 | 1263 | |||
XM_011522039.2 | XP_011520341.1 | 11 | 1359 | |||
XM_011522040.1 | XP_011520342.1 | 9 | 1208 | |||
XM_011522041.1 | XP_011520343.1 | 9 | 1208 |