Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEL1L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271539.1 | NP_001258468.1 | 9 | 893 | ||
NR_073206.1 | 10 | 1014 | ||||
NR_073207.1 | 9 | 1061 | ||||
XM_006723646.2 | XP_006723709.1 | 9 | 895 | |||
XM_006723647.2 | XP_006723710.1 | 9 | 895 | |||
XM_006723648.2 | XP_006723711.1 | 9 | 895 | |||
XM_006723650.2 | XP_006723713.1 | 9 | 895 | |||
XM_006723651.2 | XP_006723714.1 | 9 | 895 | |||
XM_006723652.2 | XP_006723715.1 | 9 | 895 | |||
XM_006723654.3 | XP_006723717.1 | 1 | 38 | |||
XM_006723655.2 | XP_006723718.1 | 9 | 895 | |||
XM_006723656.2 | XP_006723719.1 | 9 | 895 | |||
XM_011529374.1 | XP_011527676.1 | 8 | 835 | |||
XM_011529377.1 | XP_011527679.1 | 10 | 1104 | |||
XM_011529378.1 | XP_011527680.1 | 7 | 613 | |||
XM_011529379.1 | XP_011527681.1 | 7 | 646 | |||
GenBank mRNA | AK302170.1 | - | 9 | 890 | ||
AL137678.1 | - | 9 | 946 |