Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASXL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030632.2 | NP_085135.1 | 11 | 1461 | ||
XM_005258356.1 | XP_005258413.1 | 12 | 1434 | |||
XM_011526205.2 | XP_011524507.1 | 12 | 8224 | |||
XM_011526206.2 | XP_011524508.1 | 12 | 4989 | |||
XM_011526209.1 | XP_011524511.1 | 13 | 1618 | |||
XM_011526210.1 | XP_011524512.1 | 13 | 1675 | |||
XM_011526212.1 | XP_011524514.1 | 12 | 1579 | |||
XM_011526213.1 | XP_011524515.1 | 12 | 1672 | |||
XM_017026012.1 | XP_016881501.1 | 12 | 4836 | |||
XM_017026013.1 | XP_016881502.1 | 13 | 5198 | |||
XM_017026014.1 | XP_016881503.1 | 13 | 1635 | |||
GenBank mRNA | AK131454.1 | - | 13 | 1504 |