Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UNK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080419.2 | NP_001073888.2 | 12 | 1613 | ||
XM_006722145.3 | XP_006722208.1 | 15 | 5958 | |||
XM_006722148.2 | XP_006722211.1 | 13 | 1754 | |||
XM_011525400.1 | XP_011523702.1 | 13 | 1682 | |||
XM_011525401.2 | XP_011523703.1 | 7 | 770 | |||
XM_017025247.1 | XP_016880736.1 | 15 | 5994 | |||
XM_017025248.1 | XP_016880737.1 | 14 | 5896 | |||
XM_017025249.1 | XP_016880738.1 | 14 | 2039 | |||
XM_017025250.1 | XP_016880739.1 | 13 | 1941 | |||
GenBank mRNA | AB051540.1 | - | 12 | 1596 | ||
BC053362.1 | - | 12 | 1596 |