Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ROBO4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301088.1 | NP_001288017.1 | 9, 10 | 1446 | ||
NM_019055.5 | NP_061928.4 | 9, 10 | 1652 | |||
XM_006718861.1 | XP_006718924.1 | 9, 10 | 1585 | |||
XM_011542875.1 | XP_011541177.1 | 3, 4 | 414 | |||
GenBank mRNA | AF361473.1 | - | 9, 10 | 1581 | ||
AK000805.1 | - | 1, 2 | 108 | |||
AK027852.1 | - | 9, 10 | 1551 | |||
AK074163.1 | - | 8, 9 | 1182 | |||
AK289769.1 | - | 9, 10 | 1551 | |||
AK297845.1 | - | 3, 4 | 414 | |||
AK302628.1 | - | 9, 10 | 1316 | |||
AY358083.1 | - | 9, 10 | 1503 | |||
BC014995.1 | - | 8, 9 | 1264 | |||
BC039602.1 | - | 9, 10 | 1540 | 5590503 | ||
BC111562.1 | - | 9, 10 | 1546 | 40068748 | ||
BC111748.1 | - | 9, 10 | 1546 | 40068752 |