Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00152 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024204.1 | 4 | 719 | |||
NR_024205.1 | 3 | 555 | ||||
NR_024206.1 | 2 | 409 | ||||
GenBank mRNA | BC009508.2 | - | 2 | 374 | ||
BM989746.1 | - | 2 | 404 | |||
CF129206.1 | - | 2 | 447 |
Gene Symbol: MIR4435-2HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024373.2 | 2 | 495 | |||
GenBank mRNA | BC014776.1 | - | 2 | 422 | ||
BC070201.1 | - | 2 | 382 |