Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRSF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031684.2 | NP_001026854.1 | 8 | 2245 | ||
NM_001195446.1 | NP_001182375.1 | 7 | 2209 | |||
GenBank mRNA | AK091425.1 | - | 9 | 2512 | ||
AY513287.1 | - | 8 | 2091 | |||
BC022328.1 | - | 8 | 2113 | 4251314 |