Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001032391.1 | NP_001027563.1 | 7, 8 | 885 | ||
NM_016309.2 | NP_057393.2 | 9, 10 | 1050 | |||
XM_005255354.3 | XP_005255411.1 | 9, 10 | 839 | |||
XM_011545862.2 | XP_011544164.1 | 8, 9 | 999 | |||
XM_011545864.1 | XP_011544166.1 | 10, 11 | 1164 | |||
GenBank mRNA | AF037601.1 | - | 9, 10 | 926 | ||
AF151826.1 | - | 9, 10 | 1034 | |||
AK223364.1 | - | 10, 11 | 1208 | |||
AK225271.1 | - | 9, 10 | 994 | |||
AK291885.1 | - | 9, 10 | 994 | |||
AK314409.1 | - | 9, 10 | 995 | |||
AL137283.1 | - | 9, 10 | 854 | |||
AL560574.3 | - | 7, 8 | 876 | |||
BC001214.1 | - | 9, 10 | 1017 | 3533656 | ||
BC014217.2 | - | 10, 11 | 1138 | 3959936 |