Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OAF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178507.2 | NP_848602.1 | 4 | 1600 | ||
GenBank mRNA | BC047726.1 | - | 4 | 1600 | 5763917 |