Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRTAP10-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198694.3 | NP_941967.3 | 1 | 876 | ||
GenBank mRNA | AB076352.1 | - | 1 | 876 | ||
AJ566384.1 | - | 1 | 72 | |||
BC120949.1 | - | 1 | 875 | 40115143 | ||
BC120950.1 | - | 1 | 875 | 40115146 |