Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACSM2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105069.1 | NP_001098539.1 | 6 | 1000 | ||
NM_182617.3 | NP_872423.3 | 7 | 1035 | |||
XM_005255304.3 | XP_005255361.1 | 6 | 939 | |||
XM_011545824.2 | XP_011544126.1 | 5 | 821 | |||
XM_017023205.1 | XP_016878694.1 | 6 | 939 | |||
GenBank mRNA | AB073604.1 | - | 7 | 966 | ||
AK129944.1 | - | 6 | 939 | |||
AK289416.1 | - | 6 | 939 | |||
AK309601.1 | - | 6 | 1000 | |||
AY160217.1 | - | 5 | 825 |