Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006969.3 | NP_008900.3 | 4 | 2952 | ||
NR_036599.1 | 5 | 3070 | ||||
NR_036600.1 | 3 | 2825 | ||||
XM_006723354.3 | XP_006723417.1 | 4 | 2927 | |||
XM_011527262.2 | XP_011525564.1 | 4 | 2982 | |||
GenBank mRNA | AK091553.1 | - | 1 | 860 | ||
AK092385.1 | - | 1 | 1759 | |||
AK131368.1 | - | 2 | 3550 | |||
AK131574.1 | - | 1 | 2337 | |||
AK304704.1 | - | 4 | 2916 | |||
BC140720.1 | - | 3 | 2853 | 9021588 | ||
BX640767.1 | - | 1 | 1979 | |||
CR933598.1 | - | 4 | 2915 |