Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRINA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000837.1 | NP_000828.1 | 4 | 882 | ||
NM_001009184.1 | NP_001009184.1 | 4 | 770 | |||
GenBank mRNA | AB097026.1 | - | 4 | 694 | ||
AK127640.1 | - | 4 | 918 | |||
AK294127.1 | - | 5 | 627 | |||
BC041788.1 | - | 4 | 728 | 4738372 | ||
BC065834.1 | - | 3 | 636 | |||
BC084553.1 | - | 3 | 619 | 6206624 |