Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YDJC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017964.1 | NP_001017964.1 | 5 | 1112 | ||
XM_011529906.2 | XP_011528208.1 | 4 | 1209 | |||
GenBank mRNA | BC096754.1 | - | 5 | 1185 | ||
BC110075.1 | - | 4 | 918 | |||
BC110799.1 | - | 5 | 1156 | |||
BC133043.1 | - | 5 | 1266 | |||
BC133047.1 | - | 5 | 1266 | |||
BQ050386.1 | - | 1 | 81 |