Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHF19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009936.2 | NP_001009936.1 | 3, 4 | 516 | ||
NM_001286840.1 | NP_001273769.1 | 3, 4 | 387 | |||
NM_001286843.1 | NP_001273772.1 | 3, 4 | 516 | |||
NM_015651.2 | NP_056466.1 | 3, 4 | 516 | |||
XM_005251906.2 | XP_005251963.1 | 3, 4 | 324 | |||
XM_011518509.2 | XP_011516811.1 | 3, 4 | 367 | |||
XM_011518511.2 | XP_011516813.1 | 3, 4 | 850 | |||
XM_011518515.2 | XP_011516817.1 | 3, 4 | 851 | |||
XM_011518516.2 | XP_011516818.1 | 3, 4 | 853 | |||
XM_017014612.1 | XP_016870101.1 | 3, 4 | 356 | |||
XM_017014613.1 | XP_016870102.1 | 3, 4 | 854 | |||
GenBank mRNA | AK122744.1 | - | 3, 4 | 376 | ||
BC022374.1 | - | 3, 4 | 376 | |||
BC108663.1 | - | 3, 4 | 483 | |||
BC125076.1 | - | 2, 3 | 288 | 40117657 | ||
BC125077.1 | - | 2, 3 | 288 | 40117658 | ||
BX640713.1 | - | 3, 4 | 388 |