Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD18A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147195.2 | NP_671728.2 | 9 | 1409 | ||
XM_006716749.3 | XP_006716812.1 | 10 | 1686 | |||
XM_011517836.2 | XP_011516138.1 | 10 | 1687 | |||
XM_011517837.2 | XP_011516139.1 | 10 | 1687 | |||
XM_011517839.2 | XP_011516141.1 | 9 | 1631 | |||
XM_017014576.1 | XP_016870065.1 | 11 | 1818 | |||
XM_017014577.1 | XP_016870066.1 | 11 | 1811 | |||
GenBank mRNA | AB095935.1 | - | 9 | 1354 | ||
BC152434.1 | - | 9 | 1377 | 8862553 | ||
BQ787286.1 | - | 1 | 281 |
Gene Symbol: ANKRD18B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244752.1 | NP_001231681.1 | 9 | 1432 | ||
XM_011517881.2 | XP_011516183.1 | 10 | 1499 | |||
XM_011517883.2 | XP_011516185.1 | 9 | 1442 | |||
XM_011517885.2 | XP_011516187.1 | 4 | 587 | |||
XM_017014728.1 | XP_016870217.1 | 4 | 584 |