Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUFT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301317.1 | NP_001288246.1 | 3 | 298 | ||
NM_020127.2 | NP_064512.1 | 2 | 163 | |||
XM_017002223.1 | XP_016857712.1 | 2 | 239 | |||
GenBank mRNA | AF254260.1 | - | 2 | 163 | ||
AK298066.1 | - | 3 | 298 | |||
AK315414.1 | - | 2 | 159 | |||
AL136917.1 | - | 2 | 151 | |||
BC008301.1 | - | 2 | 143 | 3506004 |