Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LGALS14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020129.2 | NP_064514.1 | 3 | 342 | ||
NM_203471.1 | NP_982297.1 | 4 | 647 | |||
GenBank mRNA | AF123762.1 | - | 2 | 119 | ||
AF267852.1 | - | 3 | 342 | |||
AF367430.1 | - | 3 | 342 | |||
AF367431.1 | - | 3 | 342 | |||
AF367432.1 | - | 3 | 342 | |||
AK312040.1 | - | 3 | 181 | |||
BC022257.1 | - | 3 | 198 | 4770140 | ||
BG434228.1 | - | 4 | 473 | |||
FJ613341.1 | - | 3 | 146 | |||
FJ613342.1 | - | 3 | 146 |