Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD30B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145029.1 | NP_001138501.1 | 1 | 374 | ||
XM_006722325.3 | XP_006722388.1 | 1 | 202 | |||
XM_011525662.2 | XP_011523964.1 | 1 | 202 | |||
XM_011525664.2 | XP_011523966.1 | 1 | 202 | |||
XM_011525665.2 | XP_011523967.1 | 1 | 279 | |||
XM_011525666.2 | XP_011523968.1 | 1 | 279 | |||
GenBank mRNA | AF269088.1 | - | 1 | 374 | ||
BC028407.1 | - | 282 |