Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MGP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000900.3 | NP_000891.2 | 3 | 246 | ||
NM_001190839.1 | NP_001177768.1 | 4 | 321 | |||
GenBank mRNA | AK312029.1 | - | 3 | 181 | ||
AY542304.1 | - | 3 | 129 | |||
BC005272.1 | - | 3 | 181 | 3930143 | ||
BC070314.1 | - | 3 | 154 | 6653229 | ||
BC093078.1 | - | 3 | 181 | 30405585 | ||
BF699158.1 | - | 260 | ||||
BT006733.1 | - | 3 | 118 | |||
CB108937.1 | - | 3 | 213 | |||
CR450358.1 | - | 3 | 118 | |||
M58549.1 | - | 3 | 144 | |||
X07362.1 | - | 3 | 124 | |||
X53331.1 | - | 3 | 164 |