Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM71E1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308429.1 | NP_001295358.1 | 4 | 960 | ||
NM_138411.2 | NP_612420.1 | 4 | 912 | |||
XM_011526405.2 | XP_011524707.2 | 4 | 617 | |||
XM_011526406.2 | XP_011524708.2 | 4 | 569 | |||
GenBank mRNA | AK125745.1 | - | 4 | 467 | ||
AK131032.1 | - | 4 | 419 | |||
BC004941.1 | - | 4 | 371 | |||
BC012203.1 | - | 4 | 513 | |||
BC071737.1 | - | 4 | 912 | 5192796 |