Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDH1L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034173.3 | NP_001029345.2 | 19 | 2358 | ||
NR_027752.1 | 19 | 2354 | ||||
XM_011537986.2 | XP_011536288.1 | 17 | 1917 | |||
XM_017018889.1 | XP_016874378.1 | 20 | 2564 | |||
GenBank mRNA | AK095827.1 | - | 19 | 2253 | ||
AK300373.1 | - | 19 | 2253 | |||
BC103934.1 | - | 19 | 2253 | 40008174 | ||
BC103935.1 | - | 19 | 2249 | 40008176 | ||
CR627287.1 | - | 15 | 2222 |