Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP221 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271049.1 | NP_001257978.1 | 15 | 1629 | ||
NR_073132.1 | 16 | 1774 | ||||
NR_073133.1 | 14 | 1528 | ||||
XM_006712353.3 | XP_006712416.1 | 15 | 1669 | |||
XM_017003550.1 | XP_016859039.1 | 15 | 1669 | |||
XM_017003551.1 | XP_016859040.1 | 15 | 1796 | |||
XM_017003552.1 | XP_016859041.1 | 15 | 1682 | |||
XM_017003553.1 | XP_016859042.1 | 15 | 2015 | |||
XM_017003554.1 | XP_016859043.1 | 14 | 1511 | |||
XM_017003555.1 | XP_016859044.1 | 14 | 1511 | |||
XM_017003556.1 | XP_016859045.1 | 14 | 1514 | |||
XM_017003557.1 | XP_016859046.1 | 13 | 1444 | |||
XM_017003558.1 | XP_016859047.1 | 15 | 1669 | |||
XM_017003559.1 | XP_016859048.1 | 12 | 2640 | |||
GenBank mRNA | AK126306.1 | - | 2 | 179 | ||
AK293665.1 | - | 14 | 1511 | |||
BC036530.1 | - | 16 | 1740 | 4828213 |