Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCAF4L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK122914.1 | - | 1 | 1676 | ||
BC035027.1 | - | 1 | 1631 | 4828469 | ||
BC052943.1 | - | 1 | 1698 | 5268527 |