Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB40AL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031834.1 | NP_001027004.1 | 1 | 961 | ||
GenBank mRNA | BC101169.1 | - | 1 | 961 | 40016064 | |
BC101170.2 | - | 1 | 961 | 40016065 | ||
BC101171.1 | - | 1 | 961 | 40016067 | ||
BC101172.1 | - | 1 | 961 | 40016069 |