Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OTOR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020157.3 | NP_064542.1 | 2 | 244 | ||
XM_017027959.1 | XP_016883448.1 | 2 | 220 | |||
GenBank mRNA | AF233261.1 | - | 2 | 239 | ||
AF243505.1 | - | 2 | 202 | |||
AJ242552.1 | - | 2 | 195 | |||
AY359082.1 | - | 2 | 232 | |||
BC101688.1 | - | 2 | 244 | 8069194 | ||
BC101690.1 | - | 2 | 244 | 8069196 |