Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COX19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031617.2 | NP_001026788.1 | 3 | 912 | ||
GenBank mRNA | AK023185.1 | - | 3 | 848 | ||
AY957566.1 | - | 3 | 862 | |||
BC110420.1 | - | 3 | 849 | 5208612 |